• USA Home
  • Search Results

  • cortisone360445306511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cortisone

3 Products
Synonym:   17α,21-Dihydroxy-4-pregnene-3,11,20-trione, 17α-Hydroxy-11-dehydrocorticosterone, 4-Pregnene-17α,21-diol-3,11,20-trione, Kendall’s Compound E, Reichstein’s Substance Fa
Image of Cortisone
CAS Number:
Empirical Formula (Hill Notation):
 C21H28O5
Molecular Weight:
 360.44
 Beilstein/REAXYS Number:
 1356062
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
Show More