• USA Home
  • Search Results

  • corynoline367401879779011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Corynoline

2 Products
Synonym:   (+)-Corynoline, 13-Methylchelidonine
Image of Corynoline
CAS Number:
Empirical Formula (Hill Notation):
 C21H21NO5
Molecular Weight:
 367.40
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More