• USA Home
  • Search Results

  • coumafuryl2982911752211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Coumafuryl

1 Products
Synonym:   3-(α-Acetonyl-2-furylmethyl)-4-hydroxycoumarin, 3-(α-Acetonylfuryl)-4-hydroxy-coumarin
Image of Coumafuryl
CAS Number:
Empirical Formula (Hill Notation):
 C17H14O5
Molecular Weight:
 298.29
 Beilstein/REAXYS Number:
 6817711
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More