• USA Home
  • Search Results

  • coumarin146149164511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Coumarin

7 Products
Synonym:   1,2-Benzopyrone, 1-Benzopyran-2-one, 2H-Chromen-2-one
Image of Coumarin
CAS Number:
Empirical Formula (Hill Notation):
 C9H6O2
Molecular Weight:
 146.14
 Beilstein/REAXYS Number:
 383644
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Anal. Standard Form
  Shelf Life
  Organoleptic property
  Physical Form
  Application(s)
  Product Line
  Manufacturer designation
  Special Grade
Show More