• USA Home
  • Search Results

  • coumarin6350433821536011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Coumarin 6

2 Products
Synonym:   3-(2-Benzothiazolyl)-N,N-diethylumbelliferylamine, 3-(2-Benzothiazolyl)-7-(diethylamino)coumarin
Image of Coumarin 6
CAS Number:
Empirical Formula (Hill Notation):
 C20H18N2O2S
Molecular Weight:
 350.43
 Beilstein/REAXYS Number:
 1085798
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Absorption
  OLED Device Performance
  Orbital energy
Show More