• USA Home
  • Search Results

  • coumestrol2682247913011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Coumestrol

2 Products
Synonym:   7,12-Dihydroxycoumestan
Image of Coumestrol
CAS Number:
Empirical Formula (Hill Notation):
 C15H8O5
Molecular Weight:
 268.22
 Beilstein/REAXYS Number:
 266702
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More