• USA Home
  • Search Results

  • cprg585419979279711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

CPRG

1 Products
Synonym:   CPRG, Chlorophenol Red-β-D-galactopyranoside, Chlorophenol Red-β-D-galactoside, chlorophenol red-β-d-galactopyranoside
Image of CPRG
CAS Number:
Empirical Formula (Hill Notation):
 C25H22Cl2O10S
Molecular Weight:
 585.41
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More