• USA Home
  • Search Results

  • crotamiton2032848363611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Crotamiton

3 Products
Synonym:   N-Ethyl-N-(2-methylphenyl)-2-butenamide, N-Ethyl-o-crotonotoluidide, Crotamiton
Image of Crotamiton
CAS Number:
Linear Formula:
 CH3CH=CHCON(C2H5)C6H4CH3
Molecular Weight:
 203.28
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More