• USA Home
  • Search Results

  • crotonicacid860910793711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Crotonic acid

2 Products
Synonym:   ß-Methylacrylic acid, 2-Butenoic acid, ß-Methylacrylic acid, 2-Butenoic acid
Image of Crotonic acid
CAS Number:
Linear Formula:
 CH3CH=CHCOOH
Molecular Weight:
 86.09
 EC Index Number
 203-533-9
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More