• USA Home
  • Search Results

  • crotylalcoholmixtureofcisandtrans7211611791511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Crotyl alcohol, mixture of cis and trans

1 Products
Synonym:   2-Buten-1-ol
Image of Crotyl alcohol, mixture of <I>cis</I> and <I>trans</I>
CAS Number:
Linear Formula:
 CH3CH=CHCH2OH
Molecular Weight:
 72.11
 Beilstein/REAXYS Number:
 1361395
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More