• USA Home
  • Search Results

  • cryptochlorogenicacid3543190599711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cryptochlorogenic acid

2 Products
Synonym:   4-O-(3,4-Dihydroxycinnamoyl)-D-quinic acid, 4-O-Caffeoylquinic acid, Cryptochlorogenic acid
Image of Cryptochlorogenic acid
CAS Number:
Empirical Formula (Hill Notation):
 C16H18O9
Molecular Weight:
 354.31
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Titertest
Show More