• USA Home
  • Search Results

  • crystalviolet4079854862911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Crystal Violet

5 Products
Synonym:   Basic Violet 3, Gentian Violet, Hexamethylpararosaniline chloride, Methyl Violet 10B
Image of Crystal Violet
CAS Number:
Linear Formula:
 C25H30N3Cl
Molecular Weight:
 407.98
 Beilstein/REAXYS Number:
 3580948
 Colour Index Number:
 42555
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Mode of Action
  Density
  Restriction
  Physical Form
  Suitability
  Solubility
  Application(s)
  Activity Spectrum
  Special Grade
Show More