• USA Home
  • Search Results

  • crystalvioletsolution4079854862911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Crystal violet solution

3 Products
Image of Crystal violet solution
CAS Number:
Empirical Formula (Hill Notation):
 C25H30ClN3
Molecular Weight:
 407.98
 Beilstein/REAXYS Number:
 3580948
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Shelf Life
  pH Value
  Physical Form
  Concentration
Show More