• USA Home
  • Search Results

  • curcumin3683845837711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Curcumin

9 Products
Synonym:   (E,E)-1,7-bis(4-Hydroxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione, Diferuloylmethane, Diferulylmethane, Natural Yellow 3
Image of Curcumin
CAS Number:
Linear Formula:
 [HOC6H3(OCH3)CH=CHCO]2CH2
Molecular Weight:
 368.38
 Beilstein/REAXYS Number:
 2306965
 Colour Index Number:
 75300
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Shelf Life
  Manufacturer designation
  Melting Point
  Application(s)
  Physical Form
  Special Grade
  Solubility
Show More