• USA Home
  • Search Results

  • cyanidin35diglucosidechloride64698261167811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cyanidin 3,5-diglucoside chloride

1 Products
Synonym:   Cyanidin-3,5-di-O-glucoside, Cyanin chloride
Image of Cyanidin 3,5-diglucoside chloride
CAS Number:
Empirical Formula (Hill Notation):
 C27H31ClO16
Molecular Weight:
 646.98
 MDL number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More