• USA Home
  • Search Results

  • cyanidin3oalarabinosidechloride4548111161304811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cyanidin 3-O-α-L-arabinoside chloride

1 Products
Synonym:   Cyanidin 3-α-L-arabinopyranoside, Cyanidin 3-α-L-arabinoside, Cyanidin 3-arabinoside chloride
Image of Cyanidin 3-<I>O</I>-α-<SC>L</SC>-arabinoside chloride
CAS Number:
Empirical Formula (Hill Notation):
 C20H19ClO10
Molecular Weight:
 454.81
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More