• USA Home
  • Search Results

  • cyanoacetamide840810791511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cyanoacetamide

1 Products
Image of Cyanoacetamide
CAS Number:
Linear Formula:
 NCCH2CONH2
Molecular Weight:
 84.08
 Beilstein/REAXYS Number:
 878221
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
108448 Cyanoacetamide, 99%,