• USA Home
  • Search Results

  • cyanocobalamin1355376819911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cyanocobalamin

5 Products
Synonym:   α-(5,6-Dimethylbenzimidazolyl)cyanocobamide, CN-Cbl, Cyanocob(III)alamin, Cyanocobalamin, Cyanocobalmin, Vitamin B12, Vitamin B12
Image of Cyanocobalamin
CAS Number:
Empirical Formula (Hill Notation):
 C63H88CoN14O14P
Molecular Weight:
 1355.37
 Beilstein/REAXYS Number:
 4122889
 EC Number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Anal. Standard Form
  Manufacturer designation
  Melting Point
  Agency/Method
  Application(s)
  Biological Source
  Assay
  Color
  Physical Form
  Special Grade
Show More