• USA Home
  • Search Results

  • cyclobenzaprinehydrochloride31185620223911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cyclobenzaprine hydrochloride

4 Products
Synonym:   5-(3-Dimethylaminopropylidene)dibenzo[a,e]cycloheptatriene hydrochloride
Image of Cyclobenzaprine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C20H21N · HCl
Molecular Weight:
 311.85
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Special Grade
Show More