• USA Home
  • Search Results

  • cyclohexane841611082711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cyclohexane

16 Products
Image of Cyclohexane
CAS Number:
Empirical Formula (Hill Notation):
 C6H12
Molecular Weight:
 84.16
 Beilstein/REAXYS Number:
 1900225
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  UV-Absorption Values
  Agency/Method
  Anal. Standard Form
  Suitability
  Purity Grade
  Package quantity
  Restriction
  Refractive Index
  Product Line
  UV-Absorption Condition
  Viscosity
  Vapor pressure
  Melting Point
  Explosion limit
  Autoignition temp
  Transmittance
  Solubility
  Color
  Transition temperature
  Density
  Application(s)
  Potency
  Special Grade
  Physical Form
  Assay
Show More

Product #
Description
227048 Cyclohexane, anhydrous, 99.5%,
33117 Cyclohexane, puriss. p.a., ACS reagent, ≥99.5% (GC),
650455 Cyclohexane, for HPLC, ≥99.9%,
34855 Cyclohexane, for HPLC, ≥99.7%,
179191 Cyclohexane, ACS reagent, ≥99%,
1.09666 Cyclohexane, for analysis EMSURE® ACS,ISO,Reag. Ph Eur,
C100307 Cyclohexane, Laboratory Reagent, ≥99.8%,
PHR1687 Cyclohexane, Pharmaceutical Secondary Standard; Certified Reference Material,
1.02817 Cyclohexane, for gas chromatography ECD and FID SupraSolv®,
1.02827 Cyclohexane, for liquid chromatography LiChrosolv®,
1.02832 Cyclohexane, EMPLURA®,
676861 Cyclohexane, ACS reagent, ≥99%,
1.00667 Cyclohexane, for gas chromatography MS SupraSolv®,
1.02822 Cyclohexane, for spectroscopy Uvasol®,
RTC000089 Cyclohexane, Pharmaceutical Secondary Standard; Certified Reference Material,
1.02830 Cyclohexane, for denaturation,