• USA Home
  • Search Results

  • cyclohexanecarboxylicacid128179889511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cyclohexanecarboxylic acid

4 Products
Synonym:   Hexahydrobenzoic acid
Image of Cyclohexanecarboxylic acid
CAS Number:
Linear Formula:
 C6H11CO2H
Molecular Weight:
 128.17
 Beilstein/REAXYS Number:
 970529
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Agency/Method
  Solubility
  Special Grade
Show More