• USA Home
  • Search Results

  • cyclohexanol1001610893011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cyclohexanol

3 Products
Synonym:   Hexahydrophenol, Hexahydrophenol
Image of Cyclohexanol
CAS Number:
Empirical Formula (Hill Notation):
 C6H12O
Molecular Weight:
 100.16
 EC Index Number
 203-630-6
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Titertest
  Special Grade
Show More