• USA Home
  • Search Results

  • cyclohexanone981410894111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cyclohexanone

7 Products
Synonym:   Cyclohexanone, Pimelic ketone
Image of Cyclohexanone
CAS Number:
Empirical Formula (Hill Notation):
 C6H10O
Molecular Weight:
 98.14
 EC Index Number:
 203-631-1
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Purity Grade
  Refractive Index
  Special Grade
Show More