• USA Home
  • Search Results

  • cycloheximidesolution281356681911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cycloheximide solution

2 Products
Synonym:   CHX
Image of Cycloheximide solution
CAS Number:
Empirical Formula (Hill Notation):
 C15H23NO4
Molecular Weight:
 281.35
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Physical Form
  Concentration
Show More