• USA Home
  • Search Results

  • cyclohexylacetate1422062245711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cyclohexyl acetate

2 Products
Image of Cyclohexyl acetate
CAS Number:
Linear Formula:
 CH3COOC6H11
Molecular Weight:
 142.20
 Council of Europe no.:
 217
 EC Number:
 FEMA Number:
 2349
 Flavis number:
 9.027
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
Show More