• USA Home
  • Search Results

  • cyclooctatetraene1041562920911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cyclooctatetraene

1 Products
Image of Cyclooctatetraene
CAS Number:
Empirical Formula (Hill Notation):
 C8H8
Molecular Weight:
 104.15
 Beilstein/REAXYS Number:
 2496800
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
138924 Cyclooctatetraene, 98%,