• USA Home
  • Search Results

  • cyclopentane701328792311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cyclopentane

5 Products
Image of Cyclopentane
CAS Number:
Empirical Formula (Hill Notation):
 C5H10
Molecular Weight:
 70.13
 Beilstein/REAXYS Number:
 1900195
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Vapor pressure
  Transition temperature
  Application(s)
  Potency
  Melting Point
  Explosion limit
  Density
  Autoignition temp
  UV-Absorption Values
  Assay
  Special Grade
Show More