• USA Home
  • Search Results

  • cyclopentanethiol10220167907811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cyclopentanethiol

2 Products
Synonym:   Cyclopentyl mercaptan
Image of Cyclopentanethiol
CAS Number:
Linear Formula:
 C5H9SH
Molecular Weight:
 102.20
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Assay
Show More