• USA Home
  • Search Results

  • cyclopropanecarbonitrile6709550021011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cyclopropanecarbonitrile

1 Products
Synonym:   Cyanocyclopropane, Cyclopropyl cyanide
Image of Cyclopropanecarbonitrile
CAS Number:
Linear Formula:
 C3H5CN
Molecular Weight:
 67.09
 Beilstein/REAXYS Number:
 635790
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More