• USA Home
  • Search Results

  • cyclopropyldiphenylsulfoniumtetrafluoroborate314153346281611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cyclopropyldiphenylsulfonium tetrafluoroborate

1 Products
Image of Cyclopropyldiphenylsulfonium tetrafluoroborate
CAS Number:
Linear Formula:
 C3H5S(C6H5)2BF4
Molecular Weight:
 314.15
 Beilstein/REAXYS Number:
 4172245
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More