• USA Home
  • Search Results

  • cynarin516453096413711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cynarin

2 Products
Synonym:   (1R,3R,4S,5R)-1,3-Bis[[3-(3,4-dihydroxyphenyl)-1-oxo-2-propen-1-yl]oxy]-4,5-dihydroxycyclohexanecarboxylic acid, 1,3-Dicaffeoylquinic acid, 1,5-DQA, 1,5-Dicaffeoylquinic acid, Cynarin, Cynarine
Image of Cynarin
CAS Number:
Empirical Formula (Hill Notation):
 C25H24O12
Molecular Weight:
 516.45
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Titertest
Show More