• USA Home
  • Search Results

  • cystaminedihydrochloride225205617711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cystamine dihydrochloride

3 Products
Synonym:   2,2′-Diaminodiethyl disulfide dihydrochloride, 2,2′-Dithiobis(ethylamine) dihydrochloride, Decarboxycystine dihydrochloride
Image of Cystamine dihydrochloride
CAS Number:
Linear Formula:
 NH2CH2CH2SSCH2CH2NH2 · 2HCl
Molecular Weight:
 225.20
 Beilstein/REAXYS Number:
 3616850
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Product Line
  Melting Point
  Biological Source
  Solubility
Show More