• USA Home
  • Search Results

  • cytarabine2432214794411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cytarabine

4 Products
Synonym:   (β-D-Arabinofuranosyl)cytosine, Ara-C, Arabinocytidine, Arabinosylcytosine, Cytarabine, Cytosine β-D-arabinofuranoside, Cytosine arabinoside
Image of Cytarabine
CAS Number:
Empirical Formula (Hill Notation):
 C9H13N3O5
Molecular Weight:
 243.22
 Beilstein/REAXYS Number:
 89175
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
  Manufacturer designation
Show More