• USA Home
  • Search Results

  • cytidine243226546311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cytidine

2 Products
Synonym:   Cytosine β-D-riboside, Cytosine-1-β-D-ribofuranoside
Image of Cytidine
CAS Number:
Empirical Formula (Hill Notation):
 C9H13N3O5
Molecular Weight:
 243.22
 Beilstein/REAXYS Number:
 89173
 EC Number:
 MDL number:
 NACRES:
 NA.75
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More