• USA Home
  • Search Results

  • cytidine5triphosphatedisodiumsalt527123605168011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cytidine 5′-triphosphate disodium salt

1 Products
Synonym:   CTP
Image of Cytidine 5′-triphosphate disodium salt
CAS Number:
Empirical Formula (Hill Notation):
 C9H14N3Na2O14P3
Molecular Weight:
 527.12
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More