• USA Home
  • Search Results

  • cytochalasinafromdrechsleradematioidea477591411064611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cytochalasin A from Drechslera dematioidea

1 Products
Image of Cytochalasin A from <I>Drechslera dematioidea</I>
CAS Number:
Empirical Formula (Hill Notation):
 C29H35NO5
Molecular Weight:
 477.59
 Beilstein/REAXYS Number:
 949521
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More