• USA Home
  • Search Results

  • d2aminobutyricacid10312262391811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D-2-Aminobutyric acid

1 Products
Synonym:   (R)-(−)-2-Aminobutyric acid
Image of <SC>D</SC>-2-Aminobutyric acid
CAS Number:
Linear Formula:
 C2H5CH(NH2)CO2H
Molecular Weight:
 103.12
 Beilstein/REAXYS Number:
 1720934
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More