• USA Home
  • Search Results

  • d3vitamind261919d3solution3996798765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D3-Vitamin D2 (6,19,19-D3) solution

1 Products
Synonym:   Calciferol (6,19,19-d3) solution, Ercalciol (6,19,19-d3) solution, Ergocalciferol (6,19,19-d3) solution, Vitamin D2 (6,19,19-d3) solution
Image of D<SUB>3</SUB>-Vitamin D2 (6,19,19-D<SUB>3</SUB>) solution
Empirical Formula (Hill Notation):
 C28D3H41O
Molecular Weight:
 399.67
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Feature
Show More