• USA Home
  • Search Results

  • d9tetrahydrocannabinold3317488158639211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Δ9-Tetrahydrocannabinol-d3

1 Products
Synonym:   Δ1-Tetrahydrocannabinol-d3, 6α,7,8,10α-Tetrahydro-6,6,9-trimethyl-3-(pentyl-5,5,5-d3)-6H-dibenzo[b,d]pyran-1-ol
Image of Δ<SUP>9</SUP>-Tetrahydrocannabinol-d<SUB>3</SUB>
CAS Number:
Empirical Formula (Hill Notation):
 C21H27D3O2
Molecular Weight:
 317.48
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More