• USA Home
  • Search Results

  • d9tetrahydrocannabinolsolution31446197208311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Δ9-Tetrahydrocannabinol solution

2 Products
Synonym:   Δ1-Tetrahydrocannabinol
Image of Δ<SUP>9</SUP>-Tetrahydrocannabinol solution
CAS Number:
Empirical Formula (Hill Notation):
 C21H30O2
Molecular Weight:
 314.46
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More