• USA Home
  • Search Results

  • dDchiroinositol1801664312911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D-(+)-chiro-Inositol

1 Products
Synonym:   1,2,4/3,5,6-Hexahydroxycyclohexane
Image of <SC>D</SC>-(+)-<I>chiro</I>-Inositol
CAS Number:
Empirical Formula (Hill Notation):
 C6H12O6
Molecular Weight:
 180.16
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
468045 D-(+)-chiro-Inositol, 95%,