• USA Home
  • Search Results

  • dDgalactose180165923411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D-(+)-Galactose

6 Products
Synonym:   Galactose
Image of <SC>D</SC>-(+)-Galactose
CAS Number:
Empirical Formula (Hill Notation):
 C6H12O6
Molecular Weight:
 180.16
 Beilstein Registry Number:
 1724619
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Suitability
  pH Value
  UV-Absorption Values
  Loss
  Special Grade
  Product Line
  Solubility
Show More