• USA Home
  • Search Results

  • dDglucose180165099711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D-(+)-Glucose

12 Products
Synonym:   Dextrose
Image of <SC>D</SC>-(+)-Glucose
CAS Number:
Empirical Formula (Hill Notation):
 C6H12O6
Molecular Weight:
 180.16
 Beilstein/REAXYS Number:
 1724615
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Absorption
  Restriction
  Suitability
  UV-Absorption Values
  Quality clarifier
  Potency
  Melting Point
  Bulk Density
  Autoignition temp
  Loss
  Product Line
  Titertest
  Physical Form
  Assay
  Solubility
Show More

Product #
Description
G8270 D-(+)-Glucose, ≥99.5% (GC),
G7528 D-(+)-Glucose, BioXtra, ≥99.5% (GC),
G5767 D-(+)-Glucose, ACS reagent,
G7021 D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%,
49139 D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC),
G5146 D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma,
G6152 D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC),
47249 D-(+)-Glucose, analytical standard,
49152 D-(+)-Glucose, tested according to Ph Eur,
RDD016 D-(+)-Glucose, anhydrous, free-flowing, Redi-Dri, ≥99.5%,
1.37048 D(+)-Glucose, anhydrous EMPROVE® EXPERT Ph Eur,BP,USP,ACS,
1.08337 D(+)-Glucose, anhydrous for biochemistry Reag. Ph Eur,