• USA Home
  • Search Results

  • dDglucosesolution180165099711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D-(+)-Glucose solution

3 Products
Image of <SC>D</SC>-(+)-Glucose solution
CAS Number:
Empirical Formula (Hill Notation):
 C6H12O6
Molecular Weight:
 180.16
 Beilstein/REAXYS Number:
 1281608
 EC Number:
 MDL number:
 NACRES:
 NA.75
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Physical Form
  Application(s)
  Sterilization
  Product Line
Show More