• USA Home
  • Search Results

  • dDmalicacid1340963661311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D-(+)-Malic acid

2 Products
Synonym:   (R)-(+)-2-Hydroxysuccinic acid, D-Hydroxybutanedioic acid
Image of <SC>D-</SC>(+)-Malic acid
CAS Number:
Linear Formula:
 HO2CCH2CH(OH)CO2H
Molecular Weight:
 134.09
 Beilstein/REAXYS Number:
 1723540
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More