• USA Home
  • Search Results

  • dDmaltosemonohydrate36031636353711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D-(+)-Maltose monohydrate

6 Products
Synonym:   4-O-α-D-Glucopyranosyl-D-glucose, Maltobiose
Image of <SC>D</SC>-(+)-Maltose monohydrate
CAS Number:
Empirical Formula (Hill Notation):
 C12H22O11 · H2O
Molecular Weight:
 360.31
 Beilstein/REAXYS Number:
 93798
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  UV-Absorption Values
  Special Grade
  Product Line
  Solubility
  Melting Point
  Biological Source
  Physical Form
  Application(s)
Show More