• USA Home
  • Search Results

  • dDtrehalosedihydrate37833613823411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D-(+)-Trehalose dihydrate

5 Products
Synonym:   α,α-Trehalose, α-D-Glucopyranosyl-α-D-glucopyranoside
Image of <SC>D</SC>-(+)-Trehalose dihydrate
CAS Number:
Empirical Formula (Hill Notation):
 C12H22O11 · 2H2O
Molecular Weight:
 378.33
 Beilstein/REAXYS Number:
 5322018
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Solubility
  Product Line
  Special Grade
  Melting Point
  Physical Form
  Biological Source
  Application(s)
Show More