• USA Home
  • Search Results

  • daidzein2542448666811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Daidzein

6 Products
Synonym:   4′,7-Dihydroxy­iso­flavone, 7-Hydroxy-3-(4-hydroxy­phenyl)-4H-1-benzo­pyran-4-one, 7-Hydroxy-3-(4-hydroxy­phenyl)­chromone, Isoaurostatin
Image of Daidzein
CAS Number:
Empirical Formula (Hill Notation):
 C15H10O4
Molecular Weight:
 254.24
 Beilstein/REAXYS Number:
 231523
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Shelf Life
  Physical Form
  Application(s)
  Manufacturer designation
  Assay
Show More