• USA Home
  • Search Results

  • daidzin4163855266911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Daidzin

6 Products
Synonym:   4′,7-Dihydroxyisoflavone 7-glucoside, Daidzein 7-glucoside, Daidzein-7-O-β-D-glucopyranoside
Image of Daidzin
CAS Number:
Empirical Formula (Hill Notation):
 C21H20O9
Molecular Weight:
 416.38
 Beilstein/REAXYS Number:
 59741
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Shelf Life
  Application(s)
  Manufacturer designation
  Assay
Show More