• USA Home
  • Search Results

  • dalloisoleucine13117150935911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D-allo-Isoleucine

1 Products
Synonym:   (2R,3S)-2-Amino-3-methylpentanoic acid
Image of <SC>D</SC>-allo-Isoleucine
CAS Number:
Linear Formula:
 CH3CH2CH(CH3)CH(NH2)CO2H
Molecular Weight:
 131.17
 Beilstein/REAXYS Number:
 1721794
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More

Product #
Description
I0380 D-allo-Isoleucine,